missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD173074, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Description
PD173074 is a potent FGFR1 inhibitor with an IC50 of 25 nM and also inhibits VEGFR2 with an IC50 of 100-200 nM, showing 1000-fold selectivity for FGFR1 over PDGFR and c-Src.
Specifications
Specifications
| Chemical Name or Material | PD173074 |
| CAS | 219580-11-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H41N7O3 |
| Quantity | 200 mg |
| Solubility Information | DMSO : ≥ 52 mg/mL (99.30 mM) |
| SMILES | O=C(NC1=NC2=NC(NCCCCN(CC)CC)=NC=C2C=C1C3=CC(OC)=CC(OC)=C3)NC(C)(C)C |
| Molecular Weight (g/mol) | 523.67 |
| Formula Weight | 523.67 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction