missing translation for 'onlineSavingsMsg'
Learn More

PD168393, MedChemExpress

Product Code. 16465898
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465898 10 mg 10mg
16453396 25 mg 25mg
16452047 50 mg 50mg
16448537 5 mg 5mg
16440206 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16465898 Supplier Medchem Express Supplier No. HY13896
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PD168393 is a potent, selective and cell-permeable inhibitor of EGFR tyrosine kinase and ErbB2. PD168393 irreversiblely inactivates EGF receptor ( IC50=0.7 nM) and is inactive against insulin receptor, PDGFR, FGFR and PKC.

PD168393 is a potent, selective and cell-permeable inhibitor of EGFR tyrosine kinase and ErbB2. PD168393 irreversiblely inactivates EGF receptor ( IC50=0.7 nM) and is inactive against insulin receptor, PDGFR, FGFR and PKC.

Specifications

Chemical Name or Material PD168393
CAS 194423-15-9
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C17H13BrN4O
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (81.25 mM)
SMILES C=CC(NC1=CC2=C(NC3=CC=CC(Br)=C3)N=CN=C2C=C1)=O
Molecular Weight (g/mol) 369.22
Formula Weight 369.22
Percent Purity 98.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.