missing translation for 'onlineSavingsMsg'
Learn More

PD158780, MedChemExpress

Product Code. 16423557
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16423557 50 mg 50mg
16407538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16423557 Supplier Medchem Express Supplier No. HY18609
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PD158780 is a potent EGFR family inhibitor with IC50s of 8 pM, 49, 52, 52 nM for EGFR, ErbB2, ErbB3, and ErbB4, respectively.

PD158780 is a potent EGFR family inhibitor with IC50s of 8 pM, 49, 52, 52 nM for EGFR, ErbB2, ErbB3, and ErbB4, respectively.

Specifications

Chemical Name or Material PD158780
CAS 171179-06-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H12BrN5
Quantity 50 mg
Solubility Information DMSO : 16.67 mg/mL (50.49 mM; Need ultrasonic)
SMILES CNC1=CC2=C(NC3=CC(Br)=CC=C3)N=CN=C2C=N1
Molecular Weight (g/mol) 330.18
Formula Weight 330.18
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.