missing translation for 'onlineSavingsMsg'
Learn More

PD153035, MedChemExpress

Product Code. 16466038
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466038

Brand: Medchem Express HY14346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PD153035 (SU-5271; AG1517; ZM 252868) is a potent EGFR inhibitor with Ki and IC50 of 6 and 25 pM, respectively.

PD153035 (SU-5271

Specifications

Chemical Name or Material PD153035
CAS 153436-54-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H14BrN3O2
Quantity 10 mg
Synonym SU-5271 AG1517 ZM 252868
Solubility Information DMSO : 33.33 mg/mL (92.53 mM; Need ultrasonic)
SMILES COC1=C(C=C(C2=C1)N=CN=C2NC3=CC(Br)=CC=C3)OC
Molecular Weight (g/mol) 360.21
Formula Weight 360.21
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.