missing translation for 'onlineSavingsMsg'
Learn More

PD150606, MedChemExpress

Product Code. 16474366
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474366 100 mg 100mg
16473068 10 mg 10mg
16432606 25 mg 25mg
16425807 5 mg 5mg
16409636 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16474366 Supplier Medchem Express Supplier No. HY100529
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PD 150606 is a selective, cell-permeable non-peptide calpain inhibitor with Ki values of 0.21 μM and 0.37 μM for μ- and m-calpains respectively, which is neuroprotective.

PD 150606 is a selective, cell-permeable non-peptide calpain inhibitor with Ki values of 0.21 μM and 0.37 μM for μ- and m-calpains respectively, which is neuroprotective.

Specifications

Chemical Name or Material PD150606
CAS 179528-45-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C9H7IO2S
Quantity 100 mg
Solubility Information DMSO : ≥ 28 mg/mL (91.47 mM)
SMILES O=C(O)/C(S)=C/C1=CC=C(I)C=C1
Molecular Weight (g/mol) 306.12
Formula Weight 306.12
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.