missing translation for 'onlineSavingsMsg'
Learn More

PD0166285, MedChemExpress

Product Code. 16442067
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442067

Brand: Medchem Express HY13925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PD0166285, a substrate of P-gp, is a WEE1 inhibitor and a weak Myt1 inhibitor with IC50 values of 24 and 72 nM, respectively. PD0166285 exhibits an IC50 of 3.433 μM for Chk1.

PD0166285, a substrate of P-gp, is a WEE1 inhibitor and a weak Myt1 inhibitor with IC50 values of 24 and 72 nM, respectively. PD0166285 exhibits an IC50 of 3.433 μM for Chk1.

Specifications

Chemical Name or Material PD0166285
CAS 185039-89-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H27Cl2N5O2
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (97.57 mM; Need ultrasonic)
SMILES O=C1N(C)C2=NC(NC3=CC=C(OCCN(CC)CC)C=C3)=NC=C2C=C1C4=C(Cl)C=CC=C4Cl
Molecular Weight (g/mol) 512.43
Formula Weight 512.43
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.