missing translation for 'onlineSavingsMsg'
Learn More

Pd(NO3)2 for AAS 5g/L matrix 1% HNO3, Fisher Chemical™
BIOPHARMA

Product Code. 12926183 Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 12926183

Brand: Fisher Chemical MM 105 050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
Physical Form Liquid
Concentration 5 g/L
Type Matrix Modifier
Solvent or Matrix Nitric Acid
Quantity 50 mL
Chemical Name or Material Pd(NO3)2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.