missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L2/B7-DC/PDCD1LG2 Antibody (S02-2A1), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | PD-L2/B7-DC/PDCD1LG2 |
|---|---|
| Clone | S02-2A1 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641084
|
Novus Biologicals
NBP3-19490-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634055
|
Novus Biologicals
NBP3-19490-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal antibody specifically detects PD-L2/B7-DC/PDCD1LG2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PD-L2/B7-DC/PDCD1LG2 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
| A synthetic peptide of human PD-L2/B7-DC/PDCD1LG2 (Uniprot # Q9BQ51) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S02-2A1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 80380 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title