missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | PD-L2/B7-DC/PDCD1LG2 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 18 |
| Conjugate | DyLight 755 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human PD-L2/B7-DC/PDCD1LG2 (NP_079515.2; Met 1-Pro 219). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?