missing translation for 'onlineSavingsMsg'
Learn More

PD-L2/B7-DC/PDCD1LG2 Antibody (176605) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30678340 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30678340 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30678340 Supplier Novus Biologicals Supplier No. FAB12242J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PD-L2/B7-DC/PDCD1LG2 Monoclonal antibody specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Applications ELISA
Classification Monoclonal
Clone 176605
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PD-L2/B7-DC, Leu20-Pro219, Accession # Q9BQ51
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 80380
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.