missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Antibody (PDL1/8222R) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20876JF635
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PD-L1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Recombinant fragment (around aa190-290) of human PD-L1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
| 29126 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| PDL1/8222R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction