missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | PD-L1 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | PDL1/8222R |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fragment (around aa190-290) of human PD-L1 protein (exact sequence is proprietary) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?