missing translation for 'onlineSavingsMsg'
Learn More

PD-L1 Antibody (F2G2) - VHH - Azide and BSA Free, Novus Biologicals™

Product Code. 18338584 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338584 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338584 Supplier Novus Biologicals Supplier No. NBP312826

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Llama Monoclonal Antibody

PD-L1 Monoclonal antibody specifically detects PD-L1 in Human samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone F2G2
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS
Gene Alias Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Host Species Llama
Immunogen PD-L1 antibody was raised against a recombinant protein corresponding to amino acids 19 - 239 of human PD-L1. .
Purification Method Affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 29126
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.