missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Antibody (29E.2A3) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
PD-L1 Monoclonal specifically detects PD-L1 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry-Frozen, Functional.
Specifications
Specifications
| Antigen | PD-L1 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Functional Assay |
| Classification | Monoclonal |
| Clone | 29E.2A3 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1-4 ug/ml, Immunohistochemistry-Frozen, Functional |
| Formulation | PBS, 0.2 um filter sterilized |
| Gene Alias | Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?