missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Rabbit anti-Human, Mouse, Rat, Clone: TYMP/2890R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | PD-ECGF/Thymidine Phosphorylase |
|---|---|
| Clone | TYMP/2890R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| PD-ECGF/Thymidine Phosphorylase | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis | |
| 10 mM PBS with 0.05% BSA | |
| 1890 | |
| IgG | |
| Protein A or G purified |
| TYMP/2890R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase | |
| Recombinant full-length human PD-ECGF/Thymidine Phosphorylase (TP/ PD-ECGF) protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title