missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Mouse anti-Human, Mouse, Rat, Clone: rTYMP/3444, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07270-100ug
This item is not returnable.
View return policy
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| PD-ECGF/Thymidine Phosphorylase | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase | |
| Recombinant full-length human PD-ECGF/Thymidine Phosphorylase (TP/ PD-ECGF) protein | |
| 100 μg | |
| Angiogenesis | |
| 1890 | |
| Store at 4°C. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| rTYMP/3444 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction