missing translation for 'onlineSavingsMsg'
Learn More

PD-173952, TRC

Product Code. 30427907
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427907 10 mg
30417750 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427907 Supplier Toronto Research Chemicals Supplier No. TRCP21780010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305820-75-1
Molecular Formula C24 H21 Cl2 N5 O2
Molecular Weight (g/mol) 482.36
Synonym 6-(2,6-Dichlorophenyl)-8-methyl-2-[[4-(4-morpholinyl)phenyl]amino]pyrido[2,3-d]pyrimidin-7(8H)-one
IUPAC Name 6-(2,6-dichlorophenyl)-8-methyl-2-(4-morpholin-4-ylanilino)pyrido[2,3-d]pyrimidin-7-one
SMILES CN1C(=O)C(=Cc2cnc(Nc3ccc(cc3)N4CCOCC4)nc12)c5c(Cl)cccc5Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H21Cl2N5O2/c1-30-22-15(13-18(23(30)32)21-19(25)3-2-4-20(21)26)14-27-24(29-22)28-16-5-7-17(8-6-16)31-9-11-33-12-10-31/h2-8,13-14H,9-12H2,1H3,(H,27,28,29)
Formula Weight 481.1072
Chemical Name or Material PD-173952
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.