missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PD 168077 maleate

Product Code. 30002150
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002150 10 mg 10mg
18084692 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002150 Supplier Tocris Bioscience™ Supplier No. 1065/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity, selective D4 agonist

A potent D4 dopamine receptor agonist (Ki = 8.7 nM) with > 400-fold selectivity over D2 and > 300-fold selectivity versus D3 subtypes respectively. Induces synaptic translocation of CaMK II to postsynaptic sites in cultured prefrontal cortical neurons. Centrally active in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630117-19-0
Molecular Formula C24H26N4O5

Specifications

Quantity 10 mg
Formula Weight 450.49
Purity 98%
Molecular Weight (g/mol) 450.495
Chemical Name or Material N-[[4-(2-Cyanophenyl)-1-piperazinyl]methyl]-3-methylbenzamide maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.