missing translation for 'onlineSavingsMsg'
Learn More

PD 168077 maleate, Tocris Bioscience™

Product Code. 13488423
Encompass
Click to view available options
50mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity, selective D4 agonist

A potent D4 dopamine receptor agonist (Ki = 8.7 nM) with > 400-fold selectivity over D2 and > 300-fold selectivity versus D3 subtypes respectively. Induces synaptic translocation of CaMK II to postsynaptic sites in cultured prefrontal cortical neurons. Centrally active in vivo.

Specifications

CAS 630117-19-0
Molecular Formula C24H26N4O5
Quantity 50mg
Synonym pd 168077 maleate, pd 168,077 maleate, n-4-2-cyanophenyl piperazin-1-yl methyl-3-methylbenzamide maleate, pd 168,077 maleate salt, n-4-2-cyanophenyl piperazin-1-yl methyl-3-methylbenzamide; maleic acid, n-4-2-cyanophenyl-1-piperazinyl methyl-3-methylbenzamide maleate salt, pd168077 maleate, pd 168,077 maleate salt, powder, n-methyl-4-2-cyanophenyl piperazinyl-3-methylbenzamide maleate, n-4-2-cyanophenyl-1-piperazinyl methyl-3-methyl-benzamide maleate
InChI Key NAEUGRPISCANHO-BTJKTKAUSA-N
SMILES CC1=CC=CC(=C1)C(=O)NCN2CCN(CC2)C3=CC=CC=C3C#N.C(=CC(=O)O)C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;N-[[4-(2-cyanophenyl)piperazin-1-yl]methyl]-3-methylbenzamide
Molecular Weight (g/mol) 450.495
PubChem CID 11957665
Formula Weight 450.49
Percent Purity 98%
Chemical Name or Material N-[[4-(2-Cyanophenyl)-1-piperazinyl]methyl]-3-methylbenzamide maleate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'