missing translation for 'onlineSavingsMsg'
Learn More

PD-168077 maleate, MedChemExpress

Product Code. 16488018
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488018 10 mg 10mg
16480478 5 mg 5mg
16453736 25 mg 25mg
16437796 100 mg 100mg
16423947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16488018 Supplier Medchem Express Supplier No. HY21098A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PD-168077 maleate is a selective dopamine D4 receptor agonist, with a Ki of 9 nM.

PD-168077 maleate is a selective dopamine D4 receptor agonist, with a Ki of 9 nM.

Specifications

Chemical Name or Material PD-168077 maleate
CAS 630117-19-0
Color White
Purity Grade Notes Research
Molecular Formula C24H26N4O5
Quantity 10 mg
Solubility Information DMSO : 150 mg/mL (332.97 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NCN1CCN(C2=CC=CC=C2C#N)CC1)C3=CC=CC(C)=C3.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 450.49
Formula Weight 450.49
Percent Purity 98.74%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.