missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PD 150606

Product Code. p-4785318
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18093922

Brand: Tocris Bioscience™ 1269/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell permeable calpain inhibitor

Selective, cell-permeable non-peptide calpain inhibitor (Ki values for ν and m-calpains are 0.21 and 0.37 μM respectively). Targets the calcium binding sites of calpain. Demonstrates high specificity for calpains relative to other proteases, and is a non-competitive inhibitor with respect to the substrate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179528-45-1
Molecular Formula C9H7IO2S

Specifications

Quantity 10 mg
Formula Weight 306.12
Purity >95%
Molecular Weight (g/mol) 306.117
Chemical Name or Material PD 150606
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.