missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (+)-PD 128907 hydrochloride

Description
High affinity D3 dopamine receptor agonist (Ki = 2.3 nM). Displays 18 - 200-fold selectivity over other dopamine receptor subtypes.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 285.77 |
| Purity | >99% |
| Molecular Weight (g/mol) | 285.77 |
| Chemical Name or Material | (+)-PD 128907 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?