missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-PD 128907 hydrochloride, MedChemExpress
Description
(+)-PD 128907 hydrochloride is a selective dopamine D2/D3 receptor agonist, with Kis of 1.7, 0.84 nM for human and rat D3 receptors, 179, 770 n M for human and rat D3 receptors, respectively.
Specifications
Specifications
| Chemical Name or Material | (+)-PD 128907 hydrochloride |
| CAS | 300576-59-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H20ClNO3 |
| Quantity | 25 mg |
| Solubility Information | DMSO : 20.83 mg/mL (72.89 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (58.33 mM; Need ultrasonic) |
| SMILES | CCCN1[C@@]2([H])[C@@](OCC1)([H])C3=CC(O)=CC=C3OC2.Cl |
| Molecular Weight (g/mol) | 285.77 |
| Formula Weight | 285.77 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?