missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PD 123319 ditrifluoroacetate

Product Code. 18055692
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055692 10 mg 10mg
18024212 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18055692 Supplier Tocris Bioscience™ Supplier No. 1361/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective non-peptide AT2 antagonist

Potent, selective, non-peptide angiotensin AT2 receptor antagonist. IC50 values are 34 and 210 nM in rat adrenal tissue and brain respectively.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130663-39-7
Molecular Formula C33H33F3N4O5

Specifications

Quantity 10 mg
Formula Weight 736.67
Purity >99%
Molecular Weight (g/mol) 622.645
Chemical Name or Material PD 123319 ditrifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.