missing translation for 'onlineSavingsMsg'
Learn More

PD 123319 ditrifluoroacetate, MedChemExpress

Product Code. 16453748
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453748

Brand: Medchem Express HY10259A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PD 123319 (ditrifluoroacetate) is a potent, selective AT2 angiotensin II receptor antagonist with IC50 of 34 nM.

PD 123319 (ditrifluoroacetate) is a potent, selective AT2 angiotensin II receptor antagonist with IC50 of 34 nM.

Specifications

Chemical Name or Material PD 123319 ditrifluoroacetate
CAS 136676-91-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C35H34F6N4O7
Quantity 10 mg
Solubility Information H2O : ≥ 36 mg/mL (48.87 mM)
SMILES FC(F)(C(O)=O)F.FC(F)(C(O)=O)F.O=C([C@@H]1CC2=C(N=CN2CC3=CC=C(N(C)C)C(C)=C3)CN1C(C(C4=CC=CC=C4)C5=CC=CC=C5)=O)O
Molecular Weight (g/mol) 736.66
Formula Weight 736.66
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.