missing translation for 'onlineSavingsMsg'
Learn More

PD 117519, MedChemExpress

Product Code. 16475497
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475497 5 mg 5mg
16484216 100 mg 100mg
16469456 50 mg 50mg
16462758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475497 Supplier Medchem Express Supplier No. HY100032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PD 117519 (CI947) is an A2A adenosine agonist which has shown oral antihypertensive activity in pharmacological animal models.

PD 117519 (CI947) is an A2A adenosine agonist which has shown oral antihypertensive activity in pharmacological animal models.

Specifications

Chemical Name or Material PD 117519
CAS 96392-15-3
Color White
Purity Grade Notes Research
Molecular Formula C19H21N5O4
Quantity 5 mg
Synonym CI947
Solubility Information DMSO : ≥ 100 mg/mL (260.82 mM)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3N[C@@H]4CCC5=C4C=CC=C5)O1)O)O
Molecular Weight (g/mol) 383.4
Formula Weight 383.4
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.