missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PD-1 Recombinant Human Monoclonal Antibody (HN94)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA543078
This item is not returnable.
View return policy
Description
Specificity: Human PD1/PDCD1/CD279 KD of the antibody is <1.43E-10 M for PD1 (10377-H08H).
Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of danger signals such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
Specifications
| PD-1 | |
| Recombinant Monoclonal | |
| 5.08 mg/mL | |
| Arginine, histidine with 120mM NaCl, 0.02% Tween 80 and no preservative | |
| Q15116 | |
| Pdcd1 | |
| Recombinant Human PD1/PDCD1/CD279 Protein. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG4 |
| Neutralization | |
| HN94 | |
| Unconjugated | |
| Pdcd1 | |
| CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
| Human | |
| Protein A | |
| RUO | |
| 5133 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction