missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1/CD279 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18106-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a cell surface membrane protein of the immunoglobulin superfamily. This protein is expressed in pro-B-cells and is thought to play a role in their differentiation. In mice, expression of this gene is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure. These studies suggest that this gene product may also be important in T cell function and contribute to the prevention of autoimmune diseases.Specifications
| PD-1/CD279 | |
| Polyclonal | |
| Unconjugated | |
| PDCD1 | |
| CD279, hPD 1, hPD l, PD 1, PD1, PD-1, PDCD1, programmed cell death 1, Protein PD 1, SLEB2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18566, 5133 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02242, Q15116 | |
| PDCD1 | |
| PD-1/CD279 Fusion Protein Ag12470 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction