missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems PD-1, Biotinylated, PD-L1 Inhibitor Screening ELISA Assay Pair
Click to view available options
Quantity:
480 Tests
96 Tests
Unit Size:
480 tests
96 tests
Description
Immune checkpoint pathway is a focal point of today’s cancer research. PD-1 is one of the best characterized checkpoint proteins. The binding between PD-1 and its ligand PD-L1 suppresses T-cell activation and allows cancer cells to escape from body’s immune surveillance. Therefore, the pharmaceutical inhibition of PD-1 or its ligand has been considered a promising strategy by many oncologists.
Specifications
Specifications
| Product Type | Inhibitory Kit |
| Kit Contents | This inhibitor screening ELISA pair is designed to facilitate the identification and characterization of new PD-1 pathway inhibitors. The assay takes advantage of our in house-developed binding of biotinylated human PD-1 to immobilized human PD-L1 in a fu |
| Quantity | 96 Tests |
| Shipping Condition | RT |
| Storage Requirements | -20°C, aviod light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction