missing translation for 'onlineSavingsMsg'
Learn More

PD-1 Antibody (996209) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30732410 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732410 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732410 Supplier Novus Biologicals Supplier No. FAB77382CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

PD-1 Monoclonal antibody specifically detects PD-1 in Mouse samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-1
Applications Neutralization
Classification Monoclonal
Clone 996209
Conjugate CoraFluor 1
Formulation PBS
Gene Alias CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PD-1, Met1-Gln167, Accession # Q02242
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.