missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCYT1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531094
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, U87-MG. Predicted reactivity: Mouse (99%), Rat (99%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PCYT1B Polyclonal antibody specifically detects PCYT1B in Human samples. It is validated for Western Blot.
Specifications
| PCYT1B | |
| Polyclonal | |
| Unconjugated | |
| Pcyt1b | |
| AW045697; CCT B; CCTB; Cctbeta; CCT-beta; choline phosphate cytidylyltransferase 1 beta; choline-phosphate cytidylyltransferase B; CT B; CTB; CTP:phosphocholine cytidylyltransferase B; CTTbeta; PCYT1B; phosphate cytidylyltransferase 1, choline, beta; phosphate cytidylyltransferase 1, choline, beta isoform; phosphorylcholine transferase B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9468 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.94 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y5K3 | |
| Pcyt1b | |
| Recombinant fragment corresponding to a region within amino acids 67 and 291 of Human PCYT1B | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction