missing translation for 'onlineSavingsMsg'
Learn More

PCYT1B Antibody (F39 P7 F11), DyLight 650, Novus Biologicals™

Product Code. 18425718 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425718 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425718 Supplier Novus Biologicals Supplier No. NBP229807C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PCYT1B Monoclonal antibody specifically detects PCYT1B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCYT1B
Applications Western Blot
Classification Monoclonal
Clone F39 P7 F11
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CCT B, CCTB, CCT-betaEC 2.7.7.15, choline-phosphate cytidylyltransferase B, CT B, CTB, CTP:phosphocholine cytidylyltransferase B, EC 2.7.7, phosphate cytidylyltransferase 1, choline, beta, phosphate cytidylyltransferase 1, choline, beta isoform, Phosphorylcholine transferase B
Host Species Mouse
Immunogen Synthetic Peptide corresponding to the C-Terminus of Human CCT-beta.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 9468
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.