missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCYT1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102951
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PCYT1A.
This gene belongs to the cytidylyltransferase family and is involved in the regulation of phosphatidylcholine biosynthesis. Mutations in this gene are associated with spondylometaphyseal dysplasia with cone-rod dystrophy. Alternatively spliced transcript variants have been found for this gene.
Specifications
| PCYT1A | |
| Polyclonal | |
| Unconjugated | |
| PCYT1A | |
| CCT A; CCTA; Cctalpha; CCT-alpha; choline phosphate cytidylyltransferase 1 alpha; choline-phosphate cytidylyltransferase A; CT; CT A; CTA; CTalpha; CTP; CTP phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyltransferase A; CTP:phosphocholine cytidylyltransferase alpha; Ctpct; Cttalpha; PCYT1; PCYT1A; phosphate cytidylyltransferase 1, choline, alpha; phosphate cytidylyltransferase 1, choline, alpha isoform; phosphorylcholine transferase A; SMDCRD | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5130 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49585 | |
| PCYT1A | |
| A synthesized peptide derived from human PCYT1A(Accession P49585), corresponding to amino acid residues K307-A357. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction