missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCYT1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120510
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, HepG2, Mouse testis, Mouse brain. Immunogen sequence: MDAQCSAKVN ARKRRKEAPG PNGATEEDGV PSKVQRCAVG LRQPAPFSDE IEVDFSKPYV RVTMEEASRG TPCERPVRVY ADGIFDLFHS GHARALMQAK.
This gene belongs to the cytidylyltransferase family and is involved in the regulation of phosphatidylcholine biosynthesis. Mutations in this gene are associated with spondylometaphyseal dysplasia with cone-rod dystrophy. Alternatively spliced transcript variants have been found for this gene.
Specifications
| PCYT1A | |
| Polyclonal | |
| Unconjugated | |
| PCYT1A | |
| CCT A; CCTA; Cctalpha; CCT-alpha; choline phosphate cytidylyltransferase 1 alpha; choline-phosphate cytidylyltransferase A; CT; CT A; CTA; CTalpha; CTP; CTP phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyltransferase A; CTP:phosphocholine cytidylyltransferase alpha; Ctpct; Cttalpha; PCYT1; PCYT1A; phosphate cytidylyltransferase 1, choline, alpha; phosphate cytidylyltransferase 1, choline, alpha isoform; phosphorylcholine transferase A; SMDCRD | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13026, 5130 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P49585, P49586 | |
| PCYT1A | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PCYT1A (NP_001299602.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction