missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCYOX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557194
This item is not returnable.
View return policy
Description
Immunogen sequence: HDYAFSSVEK LLHALGGDDF LGMLNRTLLE TLQKAGFSEK FLNEMIAPVM RVNYGQSTDI NAFVGAVSLS CSDS Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 74%.
Prenylcysteine is released during the degradation of prenylated proteins. PCYOX1 catalyzes the degradation of prenylcysteine to yield free cysteines and a hydrophobic isoprenoid product (Tschantz et al., 1999 ).
Specifications
| PCYOX1 | |
| Polyclonal | |
| Unconjugated | |
| PCYOX1 | |
| 1200015P13Rik; AI115532; AI448340; chloride ion pump-associated 55 kDa protein; Clp55; KIAA0908; PCL1; Pcly; PCYOX1; Prenylcysteine lyase; prenylcysteine oxidase; Prenylcysteine oxidase 1; prenylcysteine oxidase; prenylcysteine oxidase 1; prenylcysteine oxidase-like; UNQ597/PRO1183 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51449 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UHG3 | |
| PCYOX1 | |
| Recombinant protein corresponding to Human PCYOX1. Recombinant protein control fragment (Product #RP-96038). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction