missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCTP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548782
This item is not returnable.
View return policy
Description
PCTP Polyclonal Antibody for Western Blot
Catalyzes the transfer of phosphatidylcholine between membranes. Binds a single lipid molecule. Target protein plays a role in intermembrane transfer of phosphatidylcholines.
Specifications
| PCTP | |
| Polyclonal | |
| Unconjugated | |
| PCTP | |
| PCTP; PC-TP; Phosphatidylcholine transfer protein; STARD2; StAR-related lipid transfer (START) domain containing 2; stAR-related lipid transfer protein 2; START domain-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 58488 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UKL6 | |
| PCTP | |
| KLH conjugated synthetic peptide between 59-87 amino acids from the Central region of human PCTP. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction