missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCTAIRE3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12207-1-AP-150UL
This item is not returnable.
View return policy
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway. The CMGC group consists of 60 kinases including the cyclin-dependent kinase (CDK) and close relatives family, the MAP kinase (ERK) family, the glycogen synthase kinase 3 (GSK3) family, and the Cdc2-like kinase (CLK) family.Specifications
| PCTAIRE3 | |
| Polyclonal | |
| Unconjugated | |
| CDK18 | |
| CDK18, PCTAIRE, PCTAIRE motif protein kinase 3, PCTAIRE protein kinase 3, PCTAIRE3, PCTK3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18557, 289019, 5129 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35832, Q04899, Q07002 | |
| Cdk18 | |
| PCTAIRE3 Fusion Protein Ag2851 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction