missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCTAIRE3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528326
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (92%), Rat (91%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May play a role in signal transduction cascades in terminally differentiated cells. [UniProt].
Specifications
| PCTAIRE3 | |
| Polyclonal | |
| Unconjugated | |
| Cdk18 | |
| AA682070; Cdk18; Cell division protein kinase 18; Ctk18; cyclin dependent kinase 18; cyclin-dependent kinase 18; PCTAIRE; PCTAIRE protein kinase 3; Pctaire3; PCTAIRE-motif protein kinase 3; Pctk3; Serine/threonine-protein kinase PCTAIRE-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5129 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.86 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q07002 | |
| Cdk18 | |
| Recombinant protein encompassing a sequence within the center region of human PCTAIRE3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction