missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCTAIRE1 Recombinant Rabbit Monoclonal Antibody (K01_2K72)

Product Code. 30826361
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826361 100 μL
30828048 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826361 Supplier Invitrogen™ Supplier No. MA563967

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PCTAIRE1 Recombinant Monoclonal Antibody for Western Blot

CDK16 (PCTK1)/cyclin Y belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. CDK16 plays a role in signal transduction cascades in terminally differentiated cells, in exocytosis, and in transport of secretory cargo from the endoplasmic reticulum. CDK16 is ubiquitously expressed with the highest levels detected in the brain and testis. CDK16 also contained the gene coding for ubiquitin-activating enzyme UBE1 that previously shown to mapped to Xp11.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCTAIRE1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2K72
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene CDK16
Gene Accession No. Q00536
Gene Alias Cdk16; cdk16.L; cdk16.S; Cell division protein kinase 16; Crk5; cyclin dependent kinase 16; cyclin-dependent kinase 16; cyclin-dependent kinase 16 L homeolog; cyclin-dependent kinase 16 S homeolog; FLJ16665; PCTAIRE; PCTAIRE protein kinase 1; PCTAIRE1; PCTAIRE-1 protein kinase, alternatively spliced; PCTAIRE-motif protein kinase 1; pctgaire; pctk1; PCTK1 protein; Serine/threonine-protein kinase PCTAIRE-1; testis secretory sperm-binding protein Li 224n
Gene Symbols CDK16
Host Species Rabbit
Immunogen Recombinant protein of human PCTAIRE1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5127
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.