missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCTAIRE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10102-1-AP-150UL
This item is not returnable.
View return policy
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which exhibits RNA-dependent ATPase and ATP-dependent RNA-unwinding activities. This protein is recruited to the cytoplasmic fibrils of the nuclear pore complex, where it participates in the export of mRNA from the nucleus. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| PCTAIRE1 | |
| Polyclonal | |
| Unconjugated | |
| CDK16 | |
| CDK16, PCTAIRE, PCTAIRE motif protein kinase 1, PCTAIRE protein kinase 1, PCTAIRE1, PCTGAIRE, PCTK1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18555, 5127, 81741 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q00536, Q04735, Q63686 | |
| CDK16 | |
| PCTAIRE1 Fusion Protein Ag0151 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido