missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCTAIRE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529418
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro2A, GL261. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (86%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CDK16 (PCTK1)/cyclin Y belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. CDK16 plays a role in signal transduction cascades in terminally differentiated cells, in exocytosis, and in transport of secretory cargo from the endoplasmic reticulum. CDK16 is ubiquitously expressed with the highest levels detected in the brain and testis. CDK16 also contained the gene coding for ubiquitin-activating enzyme UBE1 that previously shown to mapped to Xp11.3.
Specifications
| PCTAIRE1 | |
| Polyclonal | |
| Unconjugated | |
| CDK16 | |
| Cdk16; cdk16.L; cdk16.S; Cell division protein kinase 16; Crk5; cyclin dependent kinase 16; cyclin-dependent kinase 16; cyclin-dependent kinase 16 L homeolog; cyclin-dependent kinase 16 S homeolog; FLJ16665; PCTAIRE; PCTAIRE protein kinase 1; PCTAIRE1; PCTAIRE-1 protein kinase, alternatively spliced; PCTAIRE-motif protein kinase 1; pctgaire; pctk1; PCTK1 protein; Serine/threonine-protein kinase PCTAIRE-1; testis secretory sperm-binding protein Li 224n | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18555, 5127 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q00536, Q04735 | |
| CDK16 | |
| Recombinant protein encompassing a sequence within the center region of human PCTAIRE1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction