missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCTAIRE1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-32254CL1
This item is not returnable.
View return policy
Description
PCTAIRE1 Polyclonal antibody specifically detects PCTAIRE1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PCTAIRE1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| Cell division protein kinase 16, cyclin-dependent kinase 16, EC 2.7.11, EC 2.7.11.22, FLJ16665, PCTAIRE, PCTAIRE protein kinase 1, PCTAIRE1, PCTAIRE-motif protein kinase 1, PCTGAIRE, PCTK1Serine/threonine-protein kinase PCTAIRE-1 | |
| The immunogen this antibody was made to, maps to a region between residue 25 to 75 of human PCTAIRE-Motif Protein Kinase 1 using the numbering given in entry NP_006192.1 (GeneID 5127). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 5127 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction