missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCSK9 Recombinant Rabbit Monoclonal Antibody (HL2666)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556835
This item is not returnable.
View return policy
Description
PCSK9 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
Specifications
| PCSK9 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P59996, Q80W65, Q8NBP7 | |
| PCSK9 | |
| Recombinant fragment of human PCSK9. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2666 | |
| Unconjugated | |
| PCSK9 | |
| AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100102, 255738, 298296 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction