missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCSK9 Polyclonal Antibody
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
Specifications
Specifications
| Antigen | PCSK9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | PCSK9 |
| Gene Accession No. | Q8NBP7 |
| Gene Alias | AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9 |
| Gene Symbols | PCSK9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?