missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCSK7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12044-1-AP-150UL
This item is not returnable.
View return policy
Description
PCSK7 is likely to represent a ubiquitous endoprotease activity within constitutive secretory pathways and capable of cleavage at the RXXX[KR]R consensus motif.Specifications
| PCSK7 | |
| Polyclonal | |
| Unconjugated | |
| PCSK7 | |
| hPC8, LPC, Lymphoma proprotein convertase, PC7, PC8, PCSK7, Prohormone convertase 7, Proprotein convertase 7, Proprotein convertase 8, SPC7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18554, 29606, 9159 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16549, Q61139, Q62849 | |
| Pcsk7 | |
| PCSK7 Fusion Protein Ag2674 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction