missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCSK4 Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain.
Involved in the processing of hormone and other protein precursors at sites comprised of pairs of basic amino acid residues. Plays a role in transcriptional coactivation. May be involved in stabilizing the multiprotein transcription complex.
Specifications
Specifications
| Antigen | PCSK4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal |
| Gene | PCSK4 |
| Gene Accession No. | P29121, Q78EH2 |
| Gene Alias | AI647044; KEX2-like endoprotease 3; L21221; NEC 3; NEC3; Nec-3; Neuroendocrine convertase 3; PC4; Pcsk4; prohormone convertase 3; Proprotein convertase 4; Proprotein convertase subtilisin/kexin type 4; SPC5; testicular tissue protein Li 135; UNQ2757/PRO6496 |
| Gene Symbols | PCSK4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?