missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCSK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552168
This item is not returnable.
View return policy
Description
Immunogen sequence: ENKGYYFNTG TLYRYTLLLY GTAEDMTARP TGPQVTSSAC VQRDTEGLCQ ACDGPAYILG QLCLAYCPPR FFNHTRLVTA GPGHTAAPAL RVCSSCHASC YTCRGGSPRD CTSCPPSSTL DQQQGSCMGP T Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 35%.
Involved in the processing of hormone and other protein precursors at sites comprised of pairs of basic amino acid residues. Plays a role in transcriptional coactivation. May be involved in stabilizing the multiprotein transcription complex.
Specifications
| PCSK4 | |
| Polyclonal | |
| Unconjugated | |
| PCSK4 | |
| AI647044; KEX2-like endoprotease 3; L21221; NEC 3; NEC3; Nec-3; Neuroendocrine convertase 3; PC4; Pcsk4; prohormone convertase 3; Proprotein convertase 4; Proprotein convertase subtilisin/kexin type 4; SPC5; testicular tissue protein Li 135; UNQ2757/PRO6496 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54760 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UW60 | |
| PCSK4 | |
| Recombinant protein corresponding to Human PCSK4. Recombinant protein control fragment (Product #RP-88823). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction