missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCSK1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA529832

Product Code. 13444147

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: HepG2, HCT116. Predicted reactivity: Mouse (99%), Rat (100%), Zebrafish (93%), Pig (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a type I proinsulin-processing enzyme that plays a key role in regulating insulin biosynthesis. It is also known to cleave proopiomelanocortin, prorenin, proenkephalin, prodynorphin, prosomatostatin and progastrin. Mutations in this gene are thought to cause obesity. This encoded protein is associated with carcinoid tumors. The use of alternate polyadenylation sites has been found for this gene. Multiple alternatively spliced transcript variants have been described for this gene but their full length nature is not known.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PCSK1
Polyclonal
Unconjugated
PCSK1
Att-1; Bdp; BMIQ12; EC 3.4.21.93; Furin homolog; LOW QUALITY PROTEIN: neuroendocrine convertase 1; NEC 1; NEC1; Nec-1; Neuroendocrine convertase 1; PC1; PC3; PCSK1; Phpp-1; Prohormone convertase 1; prohormone convertase 1/3; prohormone convertase 3; propeptide-processing protease; Proprotein convertase 1; proprotein convertase subtilisin/kexin type 1; Protein convertase subtilisin / kexin type I; Protein convertase subtilisin / kexin, type I; SPC3
Rabbit
Antigen affinity chromatography
RUO
18548, 5122
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.52 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P29120, P63239
PCSK1
Recombinant protein encompassing a sequence within the center region of human PC1/3. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.