missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ PCR Tubes

Product Code. 16665472
Change view
Click to view available options
Volume (Metric):
0.2 mL
0.5 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility unitSize
16665472 0.5 mL Non-sterile Pack of 1000
16655472 0.2 mL Non-sterile Pack of 1000
16675472 0.2 mL Sterile Pack of 1000
16685472 0.5 mL Sterile Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16665472 Supplier Microspec™ Supplier No. AK2
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

PCR Tubes available in EtO sterile or non-sterile.

  • PCR tubes
  • Natural
  • Pack of 1000

Specifications

Closure Color Clear
Color Clear
Volume (Metric) 0.5 mL
Storage Requirements Room Temperature
Quantity 1000
Closure Material Polypropylene
Shape Conical
Sterility Non-sterile
Material Polypropylene
Dnase Free No
Closure Type Flip Cap
Autoclavable Not Autoclavable
Closure Included Yes
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.