missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PCR Product Pre-Sequencing Kit

Product Code. 15647748
Click to view available options
No. of Reactions:
100
2000
500 Reactions
Unit Size:
100 reactions
2000 reactions
500 reactions
This item is not returnable. View return policy

Product Code. 15647748

Brand: Applied Biosystems™ 70996.1.KT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PCR Product Pre-Sequencing Kit uses a novel, enzymatic clean-up method to pre-treat PCR products prior to sequencing, without any subsequent purification or separation steps.

  • Remove unincorporated primers and nucleotides in about 5 minutes
  • Prepare DNA sequencing samples with one pipetting step
  • Conserve precious PCR products with up to 100% recovery, regardless of amplicon length
  • Minimize spin column or bead clean-up
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5 to -30°C.
Description PCR Product Pre-Sequencing Kit, 500 rxns, Recombinant, PCR Purification, Uses a Novel, Enzymatic Clean-up Method to Pre-treat PCR Products Prior to Sequencing, without Any Subsequent Purification or Separation Steps
Format Kit
For Use With (Application) PCR Purification
No. of Reactions 500 Reactions
Product Line ExoSAP-IT
Product Type PCR Product Pre-Sequencing Kit
Protein Form Recombinant
Quantity 500 reactions
Research Category Genetic Diseases
Shipping Condition Dry Ice
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.